Merck
  • Topical v systemic agent treatment for papulopustular acne. A cost-effectiveness analysis.
Topical v systemic agent treatment for papulopustular acne. A cost-effectiveness analysis.
Archives of dermatology (1984-12-01)
R S Stern, T M Pass, A L Komaroff
ABSTRACT
We used cost-effectiveness analysis to compare the costs, risks, and benefits of two strategies for clearing papulopustular acne; topical therapy alone as initial therapy or a combination of systemic antibiotics and topical agents used at the first visit. Patients treated with combination therapy had fewer weeks of morbidity from acne, and lower dollar costs but have a greater risk of side effects. If the topical therapy alone was chosen to reduce the risk of side effects in a population of patients there would be an additional $764 in costs and 238 weeks of morbidity from acne to avert a single additional instance of side effects. Initial treatment of papulopustular acne with combined oral antibiotics and topical agents thus seems to be cost-effective. After clearing is achieved, maintenance therapy with topical agents should be considered.