Merck
  • Deletion of a Csf1r enhancer selectively impacts CSF1R expression and development of tissue macrophage populations.
Deletion of a Csf1r enhancer selectively impacts CSF1R expression and development of tissue macrophage populations.
Nature communications (2019-07-22)
Rocío Rojo, Anna Raper, Derya D Ozdemir, Lucas Lefevre, Kathleen Grabert, Evi Wollscheid-Lengeling, Barry Bradford, Melanie Caruso, Iveta Gazova, Alejandra Sánchez, Zofia M Lisowski, Joana Alves, Irene Molina-Gonzalez, Hayk Davtyan, Rebecca J Lodge, James D Glover, Robert Wallace, David A D Munro, Eyal David, Ido Amit, Véronique E Miron, Josef Priller, Stephen J Jenkins, Giles E Hardingham, Mathew Blurton-Jones, Neil A Mabbott, Kim M Summers, Peter Hohenstein, David A Hume, Clare Pridans
ABSTRACT
The proliferation, differentiation and survival of mononuclear phagocytes depend on signals from the receptor for macrophage colony-stimulating factor, CSF1R. The mammalian Csf1r locus contains a highly conserved super-enhancer, the fms-intronic regulatory element (FIRE). Here we show that genomic deletion of FIRE in mice selectively impacts CSF1R expression and tissue macrophage development in specific tissues. Deletion of FIRE ablates macrophage development from murine embryonic stem cells. Csf1rΔFIRE/ΔFIRE mice lack macrophages in the embryo, brain microglia and resident macrophages in the skin, kidney, heart and peritoneum. The homeostasis of other macrophage populations and monocytes is unaffected, but monocytes and their progenitors in bone marrow lack surface CSF1R. Finally, Csf1rΔFIRE/ΔFIRE mice are healthy and fertile without the growth, neurological or developmental abnormalities reported in Csf1r-/- rodents. Csf1rΔFIRE/ΔFIRE mice thus provide a model to explore the homeostatic, physiological and immunological functions of tissue-specific macrophage populations in adult animals.
Sigma-Aldrich
Anti-P2RY12 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-Sox9 Antibody, Chemicon®, from rabbit