Merck
  • Downregulating Neuropilin-2 Triggers a Novel Mechanism Enabling EGFR-Dependent Resistance to Oncogene-Targeted Therapies.
Downregulating Neuropilin-2 Triggers a Novel Mechanism Enabling EGFR-Dependent Resistance to Oncogene-Targeted Therapies.
Cancer research (2017-12-13)
Sabrina Rizzolio, Chiara Battistini, Gabriella Cagnoni, Maria Apicella, Viviana Vella, Silvia Giordano, Luca Tamagnone
ABSTRACT
Neuropilins are a class of cell surface proteins implicated in cell migration and angiogenesis, with aberrant expression in human tumors. Here, we show that the expression of Neuropilin-2 (NRP2) controls EGFR protein levels, thereby impinging on intracellular signaling, viability, and response to targeted therapies of oncogene-addicted cells. Notably, increased NRP2 expression in EGFR-addicted tumor cells led to downregulation of EGFR protein and tumor cell growth inhibition. NRP2 also blunted upregulation of an EGFR "rescue" pathway induced by targeted therapy in Met-addicted carcinoma cells. Cancer cells acquiring resistance to MET oncogene-targeted drugs invariably underwent NRP2 loss, a step required for EGFR upregulation. Mechanistic investigations revealed that NRP2 loss activated NFkB and upregulated the EGFR-associated protein KIAA1199/CEMIP, which is known to oppose the degradation of activated EGFR kinase. Notably, KIAA1199 silencing in oncogene-addicted tumor cells improved therapeutic responses and counteracted acquired drug resistance. Our findings define NRP2 as the pivotal switch of a novel broad-acting and actionable pathway controlling EGFR signaling, and driving resistance to therapies targeting oncogene-addiction.Significance: These important findings identify the cell surface molecule Nrp2 as the pivotal switch of a novel, actionable pathway driving EGFR upregulation and resistance to oncogene- targeted therapies. Cancer Res; 78(4); 1058-68. ©2017 AACR.
Sigma-Aldrich
Monoclonal Anti-Vinculin antibody produced in mouse, clone VIN-11-5, ascites fluid