Merck
  • Isolation and structure of a covalent cross-link adduct between mitomycin C and DNA.
Isolation and structure of a covalent cross-link adduct between mitomycin C and DNA.
Science (New York, N.Y.) (1987-03-06)
M Tomasz, R Lipman, D Chowdary, J Pawlak, G L Verdine, K Nakanishi
ABSTRACT
A DNA cross-link adduct of the antitumor agent mitomycin C (MC) to DNA has been isolated and characterized; the results provide direct proof for bifunctional alkylation of DNA by MC. Exposure of MC to Micrococcus luteus DNA under reductive conditions and subsequent nuclease digestion yielded adducts formed between MC and deoxyguanosine residues. In addition to the two known monoadducts, a bisadduct was obtained. Reductive MC activation with Na2S2O4 (sodium dithionite) leads to exclusive bifunctional alkylation. The structure of the bisadduct was determined by spectroscopic methods that included proton magnetic resonance, differential Fourier transform infrared spectroscopy, and circular dichroism. Formation of the same bisadduct in vivo was demonstrated upon injection of rats with MC. Computer-generated models of the bisadduct that was incorporated into the center of the duplex B-DNA decamer d(CGTACGTACG)2 indicated that the bisadduct fit snugly into the minor groove with minimal distortion of DNA structure. A mechanistic analysis of the factors that govern monofunctional and bifunctional adduct formation is presented.
Sigma-Aldrich
Mitomycin C ready made solution, 10 mg/mL in DMSO, ≥98% (HPLC), Cell culture tested