Merck
  • Perinatal bisphenol A (BPA) exposure alters brain oxytocin receptor (OTR) expression in a sex- and region- specific manner: A CLARITY-BPA consortium follow-up study.
Perinatal bisphenol A (BPA) exposure alters brain oxytocin receptor (OTR) expression in a sex- and region- specific manner: A CLARITY-BPA consortium follow-up study.
Neurotoxicology (2019-06-30)
Shannah K Witchey, Joelle Fuchs, Heather B Patisaul
ABSTRACT
Bisphenol A (BPA) is a well-characterized endocrine disrupting chemical (EDC) used in plastics, epoxy resins and other products. Neurodevelopmental effects of BPA exposure are a major concern with multiple rodent and human studies showing that early life BPA exposure may impact the developing brain and sexually dimorphic behaviors. The CLARITY-BPA (Consortium Linking Academic and Regulatory Insights on BPA Toxicity) program was established to assess multiple endpoints, including neural, across a wide dose range. Studies from our lab as part of (and prior to) CLARITY-BPA have shown that BPA disrupts estrogen receptor expression in the developing brain, and some evidence of oxytocin (OT) and oxytocin receptor (OTR) disruption in the hypothalamus and amygdala. While BPA disruption of steroid hormone function is well documented, less is known about its capacity to alter nonapeptide signals. In this CLARITY-BPA follow up study, we used remaining juvenile rat tissues to test the hypothesis that developmental BPA exposure affects OTR expression across the brain. Perinatal BPA exposure (2.5, 25, or 2500 μg/kg body weight (bw)/day) spanned gestation and lactation with dams gavaged from gestational day 6 until birth and then the offspring gavaged directly through weaning. Ethinyl estradiol (0.5 μg/kg bw/day) was used as a reference estrogen. Animals of both sexes were sacrificed as juveniles and OTR expression assessed by receptor binding. Our results demonstrate prenatal exposure to BPA can eliminate sex differences in OTR expression in three hypothalamic regions, and that male OTR expression may be more susceptible. Our data also identify a sub-region of the BNST with sexually dimorphic OTR expression not previously reported in juvenile rats that is also susceptible to BPA.
Sigma-Aldrich
Carboxymethylcellulose sodium salt, High viscosity