Merck
β8 Integrin Mediates Pancreatic Cancer Cell Radiochemoresistance.
Molecular cancer research : MCR (2019-07-25)
Sha Jin, Wei-Chun Lee, Daniela Aust, Christian Pilarsky, Nils Cordes
ABSTRACT
Pancreatic ductal adenocarcinoma (PDAC) stroma, composed of extracellular matrix (ECM) proteins, promotes therapy resistance and poor survival rate. Integrin-mediated cell/ECM interactions are well known to control cancer cell survival, proliferation, and therapy resistance. Here, we identified β8 integrin in a high-throughput knockdown screen in three-dimensional (3D), ECM-based cell cultures for novel focal adhesion protein targets as a critical determinant of PDAC cell radiochemoresistance. Intriguingly, β8 integrin localizes with the golgi apparatus perinuclearly in PDAC cells and resection specimen from PDAC patients. Upon radiogenic genotoxic injury, β8 integrin shows a microtubule-dependent perinuclear-to-cytoplasmic shift as well as strong changes in its proteomic interactome regarding the cell functions transport, catalysis, and binding. Parts of this interactome link β8 integrin to autophagy, which is diminished in the absence of β8 integrin. Collectively, our data reveal β8 integrin to critically coregulate PDAC cell radiochemoresistance, intracellular vesicle trafficking, and autophagy upon irradiation. IMPLICATIONS: This study identified β8 integrin as an essential determinant of PDAC cell radiochemosensitivity and as a novel potential cancer target.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Millipore
ProteoExtract® Subcellular Proteome Extraction Kit
Sigma-Aldrich
Anti-LC3B antibody, Mouse monoclonal, clone LC3B-6, purified from hybridoma cell culture
Sigma-Aldrich
Anti-APPL2 antibody produced in mouse, IgG fraction of antiserum, buffered aqueous solution