Merck
  • PID1 alters the antilipolytic action of insulin and increases lipolysis via inhibition of AKT/PKA pathway activation.
PID1 alters the antilipolytic action of insulin and increases lipolysis via inhibition of AKT/PKA pathway activation.
PloS one (2019-04-17)
Chunyan Yin, Wei Hua Liu, Yuesheng Liu, Li Wang, Yanfeng Xiao
ABSTRACT
The aim of this study was to investigate the effect of phosphotyrosine interaction domain containing 1 (PID1) on the insulin-induced activation of the AKT (protein kinase B)/protein kinase A (PKA)/hormone-sensitive lipase (HSL) pathway and lipolysis. Sprague-Dawley rats were fed either chow or a high-fat diet (HFD). The levels of insulin, glycerol, free fatty acids (FFAs) and PID1 mRNA expression were measured in the 2 groups. Furthermore, we examined the role of PID1 in the regulation of the AKT/PKA/HSL cascade and lipolysis in the 3T3-L1 cell line. Adipose tissue from HFD rats exhibited elevated PID1 expression, which showed a positive correlation with insulin levels and lipolysis. In 3T3-L1 adipocytes, we found that the antilipolytic effect of insulin is mediated by AKT and that phosphorylated AKT results in the promotion of PDE3B expression, the dephosphorylation of PKA and HSL and the suppression of glycerol release. However, overexpression of PID1 and treatment with 1 μM isoproterenol and 100 nM insulin for 24 h resulted in an increased release of glycerol and a noticeable inhibition of AKT phosphorylation, PDE3B expression and the phosphorylation of PKA/HSL in 3T3-L1 cells. In contrast, knockdown of PID1 and treatment with the above reagents inhibited lipolysis and activated the phosphorylation of AKT, which resulted in the dephosphorylation of PKA and HSL. Our findings indicate that PID1 in adipose tissue increases lipolysis by altering the antilipolytic action of insulin. This suggests that PID1 may represent a new therapeutic target to ameliorate adipocyte lipolysis and hence improve insulin sensitivity.