Merck
  • High-affinity free ubiquitin sensors for quantifying ubiquitin homeostasis and deubiquitination.
High-affinity free ubiquitin sensors for quantifying ubiquitin homeostasis and deubiquitination.
Nature methods (2019-07-17)
Yun-Seok Choi, Sarah A Bollinger, Luisa F Prada, Francesco Scavone, Tingting Yao, Robert E Cohen
ABSTRACT
Ubiquitin (Ub) conjugation is an essential post-translational modification that affects nearly all proteins in eukaryotes. The functions and mechanisms of ubiquitination are areas of extensive study, and yet the dynamics and regulation of even free (that is, unconjugated) Ub are poorly understood. A major impediment has been the lack of simple and robust techniques to quantify Ub levels in cells and to monitor Ub release from conjugates. Here, we describe avidity-based fluorescent sensors that address this need. The sensors bind specifically to free Ub, have dissociation constant Kd values down to 60 pM and, together with a newly developed workflow, allow us to distinguish and quantify the pools of free, protein-conjugated and thioesterified forms of Ub from cell lysates. Alternatively, free Ub in fixed cells can be visualized microscopically by staining with a sensor. Real-time assays using the sensors afford unprecedented flexibility and precision to measure deubiquitination of virtually any (poly)Ub conjugate.