Merck
  • Lineage-specific control of TFIIH by MITF determines transcriptional homeostasis and DNA repair.
Lineage-specific control of TFIIH by MITF determines transcriptional homeostasis and DNA repair.
Oncogene (2019-01-18)
Marcos Seoane, Sophia Buhs, Pablo Iglesias, Julia Strauss, Ann-Christin Puller, Jürgen Müller, Helwe Gerull, Susanne Feldhaus, Malik Alawi, Johanna M Brandner, Dennis Eggert, Jinyan Du, Jürgen Thomale, Peter J Wild, Martin Zimmermann, Thomas Sternsdorf, Udo Schumacher, Peter Nollau, David E Fisher, Martin A Horstmann
ABSTRACT
The melanocytic lineage, which is prominently exposed to ultraviolet radiation (UVR) and radiation-independent oxidative damage, requires specific DNA-damage response mechanisms to maintain genomic and transcriptional homeostasis. The coordinate lineage-specific regulation of intricately intertwined DNA repair and transcription is incompletely understood. Here we demonstrate that the Microphthalmia-associated transcription factor (MITF) directly controls general transcription and UVR-induced nucleotide excision repair by transactivation of GTF2H1 as a core element of TFIIH. Thus, MITF ensures the rapid resumption of transcription after completion of strand repair and maintains transcriptional output, which is indispensable for survival of the melanocytic lineage including melanoma in vitro and in vivo. Moreover, MITF controls c-MYC implicated in general transcription by transactivation of far upstream binding protein 2 (FUBP2/KSHRP), which induces c-MYC pulse regulation through TFIIH, and experimental depletion of MITF results in consecutive loss of CDK7 in the TFIIH-CAK subcomplex. Targeted for proteasomal degradation, CDK7 is dependent on transactivation by MITF or c-MYC to maintain a steady state. The dependence of TFIIH-CAK on sequence-specific MITF and c-MYC constitutes a previously unrecognized mechanism feeding into super-enhancer-driven or other oncogenic transcriptional circuitries, which supports the concept of a transcription-directed therapeutic intervention in melanoma.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-Microphthalmia (Mi) Antibody, clone C5, clone C5, Chemicon®, from mouse
Sigma-Aldrich
Anti-XPG (C-terminal) antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)