Merck
  • Untangling posterior growth and segmentation by analyzing mechanisms of axis elongation in hemichordates.
Untangling posterior growth and segmentation by analyzing mechanisms of axis elongation in hemichordates.
Proceedings of the National Academy of Sciences of the United States of America (2019-04-11)
Jens H Fritzenwanker, Kevin R Uhlinger, John Gerhart, Elena Silva, Christopher J Lowe
ABSTRACT
The trunk is a key feature of the bilaterian body plan. Despite spectacular morphological diversity in bilaterian trunk anatomies, most insights into trunk development are from segmented taxa, namely arthropods and chordates. Mechanisms of posterior axis elongation (PAE) and segmentation are tightly coupled in arthropods and vertebrates, making it challenging to differentiate between the underlying developmental mechanisms specific to each process. Investigating trunk elongation in unsegmented animals facilitates examination of mechanisms specific to PAE and provides a different perspective for testing hypotheses of bilaterian trunk evolution. Here we investigate the developmental roles of canonical Wnt and Notch signaling in the hemichordate Saccoglossus kowalevskii and reveal that both pathways play key roles in PAE immediately following the completion of gastrulation. Furthermore, our functional analysis of the role of Brachyury is supportive of a Wnt-Brachyury feedback loop during PAE in S. kowalevskii, establishing this key regulatory interaction as an ancestral feature of deuterostomes. Together, our results provide valuable data for testing hypotheses of bilaterian trunk evolution.