Merck
  • Evaluation of the effects of the solution used for electrochemical dissolution of nickel-titanium endodontic files on dentine structure, microhardness and cell viability.
Evaluation of the effects of the solution used for electrochemical dissolution of nickel-titanium endodontic files on dentine structure, microhardness and cell viability.
International endodontic journal (2018-05-16)
C C F Amaral, F Ormiga, L C Boldrini, P G Miranda, T A Mendonça, J M Granjeiro, J A C P Gomes
ABSTRACT
To evaluate the effects of the [NaF 12 g L-1  + NaCl 1 g L-1 ] solution used in the electrochemical dissolution process of fractured endodontic files, as well as its NiTi-containing product, on dentine hardness, topography and human fibroblast viability. Sixty single-rooted human teeth were evaluated for dentine microhardness using the Vickers hardness test and the area and number of dentinal tubules by scanning electron microscopy. The samples were divided according to the dentine surface treatment: distilled water; 17% EDTA; [NaF 12 g L-1  + NaCl 1 g L-1 ]; and 17% EDTA + [NaF 12 g L-1  + NaCl 1 g L-1 ]. Thirty-six single-rooted human teeth were divided according to the irrigation protocol: Dulbecco's Modified Eagle's Medium + 10% foetal bovine serum; 5.25% NaOCl; [NaF 12 g L-1  + NaCl 1 g L-1 ]; and [NaF 12 g L-1  + NaCl 1 g L-1  + NiTi]. The extracts in contact with the apical foramen were used in the MTT assay to evaluate human fibroblast viability, with dilutions of 100%, 50%, 25% and 12.5%. Statistical tests used were paired t-tests, one-way anova, Tukey's test, Kruskal-Wallis test and Dunn's post-test. The [NaF 12 g L-1  + NaCl 1 g L-1 ] solution did not modify dentine microhardness or the average dentinal tubule area. However, EDTA induced changes in dentine structure and microhardness (P < 0.05). The [NaF 12 g L-1  + NaCl 1 g L-1 ] solution, and its NiTi-containing product had lower cytotoxicity than NaOCl at dilutions of 25% and 50% (P < 0.01). The [NaF 12 g L-1  + NaCl 1 g L-1 ] solution did not alter dentine microhardness or damage the dentine structure. It also demonstrated lower cytotoxicity than NaOCl.