- Thermal constraints on in vivo optogenetic manipulations.
Thermal constraints on in vivo optogenetic manipulations.
Nature neuroscience (2019-06-19)
Scott F Owen, Max H Liu, Anatol C Kreitzer
PMID31209378
ABSTRACT
A key assumption of optogenetics is that light only affects opsin-expressing neurons. However, illumination invariably heats tissue, and many physiological processes are temperature-sensitive. Commonly used illumination protocols increased the temperature by 0.2-2 °C and suppressed spiking in multiple brain regions. In the striatum, light delivery activated an inwardly rectifying potassium conductance and biased rotational behavior. Thus, careful consideration of light-delivery parameters is required, as even modest intracranial heating can confound interpretation of optogenetic experiments.