Merck
  • Colonization and growth of dehalorespiring biofilms on carbonaceous sorptive amendments.
Colonization and growth of dehalorespiring biofilms on carbonaceous sorptive amendments.
Biofouling (2019-02-23)
Staci L Capozzi, Coline Bodenreider, Ana Prieto, Rayford B Payne, Kevin R Sowers, Birthe Veno Kjellerup
ABSTRACT
Removal of polychlorinated biphenyls (PCBs) from contaminated sediments is a priority due to accumulation in the food chain. Recent success with reduction of PCB bioavailability due to adsorption onto activated carbon led to the recognition of in situ treatment as a remediation approach. In this study, reduced bioavailability and subsequent break-down of PCBs in dehalorespiring biofilms was investigated using Dehalobium chlorocoercia DF1. DF1 formed a patchy biofilm ranging in thickness from 3.9 to 6.7 µm (average 4.6 ± 0.87 µm), while the biofilm coverage varied from 5.5% (sand) to 20.2% (activated carbon), indicating a preference for sorptive materials. Quantification of DF1 biofilm bacteria showed 1.2-15.3 × 109 bacteria per gram of material. After 22 days, coal activated carbon, bone biochar, polyoxymethylene, and sand microcosms had dechlorinated 73%, 93%, 100%, and 83%, respectively. These results show that a biofilm-based inoculum for bioaugmentation of PCBs in sediment can be an efficient approach.