- Exercise-Induced Changes to the Macrophage Response in the Dorsal Root Ganglia Prevent Neuropathic Pain after Spinal Cord Injury.
Exercise-Induced Changes to the Macrophage Response in the Dorsal Root Ganglia Prevent Neuropathic Pain after Spinal Cord Injury.
Spinal cord injury (SCI) induces neuropathic pain that is refractory to treatment. Central and peripheral immune responses to SCI play critical roles in pain development. Although immune responses in the dorsal horn have been implicated in SCI-pain, immune mechanisms in the periphery, especially in the dorsal root ganglia (DRG), where nociceptor cell bodies reside, have not been well studied. Exercise is an immunomodulator, and we showed previously that early exercise after SCI reduces pain development. However, the mechanisms of exercise-mediated pain reduction are not understood. Therefore, we examined the 1) underlying immune differences in the spinal cord and DRG between rats with and without pain and 2) immunomodulatory effects of exercise in pain reduction. Rats were subjected to a unilateral contusion at C5 and tested for pain development using von Frey and mechanical conflict-avoidance paradigms. A subgroup of rats was exercised on forced running wheels starting at 5 days post-injury for 4 weeks. We observed greater microglial activation in the C7-C8 dorsal horn of rats with SCI-induced pain compared to rats with normal sensation, and early exercise reduced this activation independently of pain behavior. Further, abnormal pain sensation strongly correlated with an increased number of DRG macrophages. Importantly, exercise-treated rats that maintain normal sensation also have a lower number of macrophages in the DRG. Our data suggest that macrophage presence in the DRG may be an important effector of pain development, and early wheel walking exercise may mediate pain prevention by modulating the injury-induced macrophage response in the DRG. Further supportive evidence demonstrated that rats that developed pain despite exercise intervention still displayed a significantly elevated number of macrophages in the DRG. Collectively, these data suggest that macrophage presence in the DRG may be an amenable cellular target for future therapies.