Merck
  • Agouti-Related Protein 2 Is a New Player in the Teleost Stress Response System.
Agouti-Related Protein 2 Is a New Player in the Teleost Stress Response System.
Current biology : CB (2019-06-11)
Inbal Shainer, Maximilian Michel, Gregory D Marquart, Ashwin A Bhandiwad, Nilli Zmora, Zohar Ben-Moshe Livne, Yonathan Zohar, Adi Hazak, Yael Mazon, Dominique Förster, Lian Hollander-Cohen, Roger D Cone, Harold A Burgess, Yoav Gothilf
ABSTRACT
Agouti-related protein (AgRP) is a hypothalamic regulator of food consumption in mammals. However, AgRP has also been detected in circulation, but a possible endocrine role has not been examined. Zebrafish possess two agrp genes: hypothalamically expressed agrp1, considered functionally equivalent to the single mammalian agrp, and agrp2, which is expressed in pre-optic neurons and uncharacterized pineal gland cells and whose function is not well understood. By ablation of AgRP1-expressing neurons and knockout of the agrp1 gene, we show that AgRP1 stimulates food consumption in the zebrafish larvae. Single-cell sequencing of pineal agrp2-expressing cells revealed molecular resemblance to retinal-pigment epithelium cells, and anatomic analysis shows that these cells secrete peptides, possibly into the cerebrospinal fluid. Additionally, based on AgRP2 peptide localization and gene knockout analysis, we demonstrate that pre-optic AgRP2 is a neuroendocrine regulator of the stress axis that reduces cortisol secretion. We therefore suggest that the ancestral role of AgRP was functionally partitioned in zebrafish by the two AgRPs, with AgRP1 centrally regulating food consumption and AgRP2 acting as a neuroendocrine factor regulating the stress axis.
Product Description
Roche
Anti-Digoxigenin-AP, Fab fragments, from sheep