Merck
  • Neuromodulatory Activity of Dietary Phenolics Derived from Corchorus olitorius L.
Neuromodulatory Activity of Dietary Phenolics Derived from Corchorus olitorius L.
Journal of food science (2019-04-25)
Reham Wagdy, Reham M Abdelkader, Ahmed H El-Khatib, Michael W Linscheid, Nabila Hamdi, Heba Handoussa
ABSTRACT
Dietary phenolics are known for their potent antioxidant and anti-inflammatory activities, making them promising candidates for protection against neuroinflammation and neurodegeneration. Hydroalcohol extract of Egyptian species of Corchorus olitorius L. (Co) leaves was investigated for its neuroprotective effects in a lipopolysaccharide-induced neuroinflammatory mouse model. Twenty five metabolites were characterized from the bioactive extract using high-performance liquid chromatography HPLC/PDA/HRESI/MSn , revealing 1,5-dicaffeoylquinic acid (Co11) as one of the major constituents (5.7%), which was isolated and its identity was confirmed by spectral data as first report. Co significantly protected microglia against H2 O2 -induced cytotoxicity and immunohistochemistry showed reduced expression of the astrocytic marker, glial fibrillary acidic protein, and the inflammatory marker, cyclooxygenase-2. These findings correlated with significant improvement of cognitive functions and reduction of LPS-induced neurodegeneration in Co-treated mice as revealed by histopathology. The current study shows promising effects of Co in limiting neurodegeneration and cognitive impairment caused by neuroinflammation and glial cell activation. PRACTICAL APPLICATION: Information presented here shed light on the promising effects of Corchorus olitorius (Co) for the modulation of neuroinflammatory pathways improving the neuroinflammation-related neurodegeneration and cognitive decline. This makes Co a promising candidate as a nutraceutical supplement to be used against neuroinflammation-related disorders.