Merck
  • Circular dichroism analysis of ligand-induced conformational changes in protein kinase C. Mechanism of translocation of the enzyme from the cytosol to the membranes and its implications.
Circular dichroism analysis of ligand-induced conformational changes in protein kinase C. Mechanism of translocation of the enzyme from the cytosol to the membranes and its implications.
The Biochemical journal (1993-03-15)
L Boscá, F Morán
ABSTRACT
The structural changes following the binding to protein kinase C (PKC) of activators that promote its translocation to lipid environments were studied by far-u.v. c.d. and intrinsic fluorescence measurements of the protein. In the absence of activators, PKC contained 40% alpha-helix, with an average size of 13 amino acids per alpha-helix segment, and 12% beta-structure as deduced from c.d. spectral analysis while fitting a set of model proteins of known structure. Ligands that promote translocation and activation of the enzyme, such as Ca2+ ions and phorbol esters, produced drastic changes in the c.d. spectra which may be interpreted as a reduction in the average number of consecutive amino acids in the alpha-helix. Most of the total alpha-helix structure was conserved and an increase in beta-structure was produced by active phorbol esters. These activators differentially affected the fluorescence of PKC: phorbol esters shifted the emission maximum to the red, whereas Ca2+ produced a marked decrease in the intensity of the fluorescence emission, suggesting in both cases that tryptophan residues were exposed to increased polar environments after binding of the ligands.