- GDP beta S enhances the activation of phospholipase C caused by thrombin in human platelets: evidence for involvement of an inhibitory GTP-binding protein.
GDP beta S enhances the activation of phospholipase C caused by thrombin in human platelets: evidence for involvement of an inhibitory GTP-binding protein.
Guanosine 5'-O-thiotriphosphate (GTP gamma S) and thrombin stimulate the activity of phospholipase C in platelets that have been permeabilized with saponin and whose inositol phospholipids have been prelabeled with [3H]inositol. Ca2+ has opposite effects on the formation of [3H]inositol phosphates induced by thrombin or GTP gamma S. While the action of GTP gamma S on the formation of [3H]inositol phosphates is inhibited by Ca2+, action of thrombin is stimulated by Ca2+. Guanosine 5'-O-(2-thiodiphosphate) (GDP beta S), which inhibits the function of GTP-binding proteins, also inhibits the effect of GTP gamma S on phospholipase C stimulation but, surprisingly, increases the effect of thrombin. Ca2+ increases the inhibitory effect of GDP beta S on GTP gamma S activation of phospholipase C, but Ca2+ further enhances the stimulatory effect of GDP beta S on the thrombin activation of phospholipase C. This indicates that two mechanisms are responsible for the activation of phospholipase C in platelets. A GTP-binding protein is responsible for regulation of phospholipase C induced by GTP gamma S, while the effect of thrombin on the stimulation of phospholipase C is independent of GTP-binding proteins. However, the effect of thrombin may be modulated by the action of an inhibitory GTP-binding protein.