Merck
  • Differential expression of thymosin beta-10 by early passage and senescent vascular endothelium is modulated by VPF/VEGF: evidence for senescent endothelial cells in vivo at sites of atherosclerosis.
Differential expression of thymosin beta-10 by early passage and senescent vascular endothelium is modulated by VPF/VEGF: evidence for senescent endothelial cells in vivo at sites of atherosclerosis.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2001-02-07)
E Vasile, Y Tomita, L F Brown, O Kocher, H F Dvorak
ABSTRACT
VPF/VEGF acts selectively on the vascular endothelium to enhance permeability, induce cell migration and division, and delay replicative senescence. To understand the changes in gene expression during endothelial senescence, we investigated genes that were differentially expressed in early vs. late passage (senescent) human dermal endothelial cells (HDMEC) using cDNA array hybridization. Early passage HDMEC cultured with or without VPF/VEGF overexpressed 9 and underexpressed 6 genes in comparison with their senescent counterparts. Thymosin beta-10 expression was modulated by VPF/VEGF and was strikingly down-regulated in senescent EC. The beta-thymosins are actin G-sequestering peptides that regulate actin dynamics and are overexpressed in neoplastic transformation. We have also identified senescent EC in the human aorta at sites overlying atherosclerotic plaques. These EC expressed senescence-associated neutral beta-galactosidase and, in contrast to adventitial microvessel endothelium, exhibited weak staining for thymosin beta-10. ISH performed on human malignant tumors revealed strong thymosin beta-10 expression in tumor blood vessels. This is the first report that Tbeta-10 expression is significantly reduced in senescent EC, that VPF/VEGF modulates thymosin beta-10 expression, and that EC can become senescent in vivo. The reduced expression of thymosin beta-10 may contribute to the senescent phenotype by reducing EC plasticity and thus impairing their response to migratory stimuli.
Sigma-Aldrich
Cellular Senescence Assay, Cellular Senescence Assay Kits provide all the reagents required to efficiently detect SA-β-gal activity at pH 6.0 in cultured cells & tissue sections.
Pricing and availability is not currently available.