Merck
  • Immunomagnetic separation and quantification of butyrylcholinesterase nerve agent adducts in human serum.
Immunomagnetic separation and quantification of butyrylcholinesterase nerve agent adducts in human serum.
Analytical chemistry (2010-07-14)
Jennifer L S Sporty, Sharon W Lemire, Edward M Jakubowski, Julie A Renner, Ronald A Evans, Robert F Williams, Jurgen G Schmidt, Marcel J van der Schans, Daan Noort, Rudolph C Johnson
ABSTRACT
A novel method for extracting butyrylcholinesterase (BuChE) from serum as a means of identifying and measuring nerve agent adducts to human BuChE is presented here. Antibutyrylcholinesterase monoclonal antibodies were conjugated to protein-G ferromagnetic particles and mixed with 500 microL serum samples. The particle-antibody-BuChE product was rinsed and directly digested with pepsin. Native and isotopically enriched nonapeptides corresponding to the pepsin digest products for uninhibited BuChE, and sarin, cyclohexylsarin, VX, and Russian VX nerve agent-inhibited BuChE were synthesized for use as calibrators and internal standards, respectively. Internal standards were added to the filtered digest sample, and the samples were quantified via high performance liquid chromatography-isotope dilution-tandem mass spectrometry. The ratio of adducted to total BuChE nonapeptides was calculated for each nerve agent-exposed serum sample using data collected in a single chromatogram. Nerve agent-inhibited quality control serum pools were characterized as part of method validation; the method was observed to have extremely low background noise. The measurement of both uninhibited and inhibited BuChE peptides compensated for any variations in the pepsin digestion before the internal standard peptide was added to the sample and may prove useful in individualizing patient results following a nerve agent exposure.
Sigma-Aldrich
Phosphate Buffered Saline, 10× PBS for Western blots and IP
Pricing and availability is not currently available.