MilliporeSigma
The effect of fenoldopam on the blood pressure of the rat.
General pharmacology (1999-01-15)
M Quevedo, J C Prieto, J Perez-Olea
ABSTRACT
1. Fenoldopam mesylate, a benzazepine derivative, is a D1 receptor agonist that lowers blood pressure through vasodilation of renal, mesenteric, coronary and cerebral vascular beds. 2. Experiments were performed in rats, and mean carotid blood pressure and heart rate were registered. Two series of experiments were performed: (1) fenoldopam as control group and (2) fenoldopam after pretreatment with one of the following drugs: the D1 antagonist SCH 23390, the D2 antagonist sulpiride, the selective beta1-adrenergic antagonist atenolol, the selective beta2-adrenergic antagonist ICI 118.551, the nonselective beta-adrenergic antagonist propranolol and the neurotoxin that destroys catecholaminergic nerve terminals 6-hydroxydopamine (6-OH-DA). 3. Fenoldopam produced a dose-dependent hypotensive effect that was not modified by pretreatment of the rat with atenolol or propranolol; however, ICI 118.551 produced a significant reduction of the hypotensive response induced by fenoldopam. 4. Pretreatment of the animals with SCH 23390 produced a significant dose-related reversal of the rat blood pressure reduction induced by low doses of fenoldopam. Sulpiride produced a result similar to that induced by pretreatment with SCH 23390. 5. The pretreatment of the animals with 6-OH-DA surprisingly attenuated the response induced by fenoldopam and produced only a significant reversal of the reduction of mean blood pressure induced with the lower dose of fenoldopam. 6. The findings obtained in the present work do not provide further evidence of direct participation of beta2-adrenergic receptors on the mechanism of action of fenoldopam. Its action seems to be mainly due to activation of D1 cardiovascular receptors.