Merck
  • Preventing the calorie restriction-induced increase in insulin-stimulated Akt2 phosphorylation eliminates calorie restriction's effect on glucose uptake in skeletal muscle.
Preventing the calorie restriction-induced increase in insulin-stimulated Akt2 phosphorylation eliminates calorie restriction's effect on glucose uptake in skeletal muscle.
Biochimica et biophysica acta (2012-08-01)
Naveen Sharma, Edward B Arias, Donel A Sequea, Gregory D Cartee
ABSTRACT
Calorie restriction (CR; ~60% of ad libitum, AL, consumption) improves insulin-stimulated glucose uptake in skeletal muscle. The precise cellular mechanism for this healthful outcome is unknown, but it is accompanied by enhanced insulin-stimulated activation of Akt. Previous research using Akt2-null mice demonstrated that Akt2 is essential for the full CR-effect on insulin-stimulated glucose uptake by muscle. However, because Akt2-null mice were completely deficient in Akt2 in every cell throughout life, it would be valuable to assess the efficacy of transient, muscle-specific Akt inhibition for attenuation of CR-effects on glucose uptake. Accordingly, we used a selective Akt inhibitor (MK-2206) to eliminate the CR-induced elevation in insulin-stimulated Akt2 phosphorylation and determined the effects on Akt substrates and glucose uptake. We incubated isolated epitrochlearis muscles from 9-month-old AL and CR (~60-65% of AL intake for 6months) rats with or without MK-2206 and measured insulin-stimulated (1.2nM) glucose uptake and phosphorylation of the insulin receptor (Tyr1162/1163), pan-Akt (Thr308 and Ser473), Akt2 (Thr308 and Ser473), AS160/TBC1D4 (Thr642), and Filamin C (Ser2213). Incubation of isolated skeletal muscles with a dose of a selective Akt inhibitor that eliminated the CR-induced increases in Akt2 phosphorylation prevented CR's effects on insulin-stimulated glucose uptake, pAS160(Thr642) and pFilamin C(Ser2213) without altering pIR(Tyr1162/1163). These data provide compelling new evidence linking the CR-induced increase in insulin-stimulated Akt2 phosphorylation to CR's effects on insulin-mediated phosphorylation of Akt substrates and glucose uptake in skeletal muscle.
Sigma-Aldrich
Anti-Glucose Transporter GLUT-4 Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-AS160 (Rab-GAP) Antibody, serum, Upstate®