Merck
  • Acetylation of snail modulates the cytokinome of cancer cells to enhance the recruitment of macrophages.
Acetylation of snail modulates the cytokinome of cancer cells to enhance the recruitment of macrophages.
Cancer cell (2014-10-15)
Dennis Shin-Shian Hsu, Hsiao-Jung Wang, Shyh-Kuan Tai, Chun-Hung Chou, Chia-Hsin Hsieh, Po-Hsien Chiu, Nien-Jung Chen, Muh-Hwa Yang
ABSTRACT
Snail is primarily known as a transcriptional repressor that induces epithelial-mesenchymal transition by suppressing adherent proteins. Emerging evidence suggests that Snail can act as an activator; however, the mechanism and biological significance are unclear. Here, we found that CREB-binding protein (CBP) is the critical factor in Snail-mediated target gene transactivation. CBP interacts with Snail and acetylates Snail at lysine 146 and lysine 187, which prevents the repressor complex formation. We further identified several Snail-activated targets, including TNF-α, which is also the upstream signal for Snail acetylation, and CCL2 and CCL5, which promote the recruitment of tumor-associated macrophages. Here, we present our results on the mechanism by which Snail induces target gene transactivation to remodel the tumor microenvironment.
Sigma-Aldrich
Acetyl coenzyme A lithium salt, ≥93% (HPLC)
Sigma-Aldrich
p300, HAT Domain, Recombinant GST-fusion protein corresponding to amino acids 1066-1707 of human p300.