Merck
  • N-Hydroxyindole-based inhibitors of lactate dehydrogenase against cancer cell proliferation.
N-Hydroxyindole-based inhibitors of lactate dehydrogenase against cancer cell proliferation.
European journal of medicinal chemistry (2011-09-29)
Carlotta Granchi, Sarabindu Roy, Alessio De Simone, Irene Salvetti, Tiziano Tuccinardi, Adriano Martinelli, Marco Macchia, Mario Lanza, Laura Betti, Gino Giannaccini, Antonio Lucacchini, Elisa Giovannetti, Rocco Sciarrillo, Godefridus J Peters, Filippo Minutolo
ABSTRACT
Current cancer research is being increasingly focused on the study of distinctive characters of tumour metabolism, resulting in a switch from oxidative phosphorylation to glycolysis (Warburg effect). Isoform 5 of human lactate dehydrogenase (hLDH5), which catalyzes the final step in the glycolytic cascade (pyruvate to lactate), constitutes a relatively new and untapped anti-cancer target. In this study, careful design and synthesis of a selected series of aryl-substituted N-hydroxyindole-2-carboxylates (NHIs) has led to several hLDH5-inhibitors, showing "first-in-class" potency and isoform selectivity. Enzyme kinetics studies indicated that these inhibitors exhibit a competitive mode of inhibition. Some representative examples were tested against two human pancreatic carcinoma cell lines, and displayed a good anti-proliferative activity, which was even more evident under hypoxic conditions.
Sigma-Aldrich
Phenylboronic acid, purum, ≥97.0% (HPLC)
Sigma-Aldrich
Phenylboronic acid, 95%