- Studies on the properties of some peripheral MAO inhibitors.
Studies on the properties of some peripheral MAO inhibitors.
Progress in neuro-psychopharmacology & biological psychiatry (1991-01-01)
E Scarr, P H Yu, B A Davis, A A Boulton
PMID1871332
ABSTRACT
1. Studies were carried out on three monoamine oxidase (MAO) inhibitors, two of which, debrisoquine and para- hydroxyphenelzine, are purported to be peripheral inhibitors and one, phenelzine, is a peripherally acting inhibitor, which has been included for comparitive purposes. 2. All three showed varying degrees of specificity towards MAO type A. 3. The action of debrisoquine was very rapid as was that of para- hydroxyphenelzine. 4. The inhibition caused by debrisoquine was competitive and reversible, while that caused by both phenelzine and para- hydroxyphenelzine was irreversible. 5. The inhibition caused by debrisoquine appeared to be unaffected by the pH of the medium.
MATERIALS