Merck
  • Integrative analysis of transcriptomics and clinical data uncovers the tumor-suppressive activity of MITF in prostate cancer.
Integrative analysis of transcriptomics and clinical data uncovers the tumor-suppressive activity of MITF in prostate cancer.
Cell death & disease (2018-10-13)
Lorea Valcarcel-Jimenez, Alice Macchia, Natalia Martín-Martín, Ana Rosa Cortazar, Ariane Schaub-Clerigué, Mikel Pujana-Vaquerizo, Sonia Fernández-Ruiz, Isabel Lacasa-Viscasillas, Aida Santos-Martin, Ana Loizaga-Iriarte, Miguel Unda-Urzaiz, Ivana Hermanova, Ianire Astobiza, Mariona Graupera, Julia Starkova, James Sutherland, Rosa Barrio, Ana M Aransay, Arkaitz Carracedo, Verónica Torrano
ABSTRACT
The dysregulation of gene expression is an enabling hallmark of cancer. Computational analysis of transcriptomics data from human cancer specimens, complemented with exhaustive clinical annotation, provides an opportunity to identify core regulators of the tumorigenic process. Here we exploit well-annotated clinical datasets of prostate cancer for the discovery of transcriptional regulators relevant to prostate cancer. Following this rationale, we identify Microphthalmia-associated transcription factor (MITF) as a prostate tumor suppressor among a subset of transcription factors. Importantly, we further interrogate transcriptomics and clinical data to refine MITF perturbation-based empirical assays and unveil Crystallin Alpha B (CRYAB) as an unprecedented direct target of the transcription factor that is, at least in part, responsible for its tumor-suppressive activity in prostate cancer. This evidence was supported by the enhanced prognostic potential of a signature based on the concomitant alteration of MITF and CRYAB in prostate cancer patients. In sum, our study provides proof-of-concept evidence of the potential of the bioinformatics screen of publicly available cancer patient databases as discovery platforms, and demonstrates that the MITF-CRYAB axis controls prostate cancer biology.
Sigma-Aldrich
5-Bromo-2′-deoxyuridine, ≥99% (HPLC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
MISSION® esiRNA, targeting human CRYAB