Merck
  • Coupling of Sulfoxonium Ylides with Arynes: A Direct Synthesis of Pro-Chiral Aryl Ketosulfoxonium Ylides and Its Application in the Preparation of α-Aryl Ketones.
Coupling of Sulfoxonium Ylides with Arynes: A Direct Synthesis of Pro-Chiral Aryl Ketosulfoxonium Ylides and Its Application in the Preparation of α-Aryl Ketones.
Organic letters (2018-10-27)
Alexánder Garay Talero, Bruna Simões Martins, Antonio C B Burtoloso
ABSTRACT
A general, mild, and versatile synthesis of the challenging α-aryl-β-ketosulfoxonium ylides has been developed for the first time, substituting traditional methods starting from diazo compounds. The arylation of easily accessible β-ketosulfoxonium ylides using aryne chemistry allowed the preparation of a large scope of the pro-chiral ylides in very good yields (40 examples; up to 85%). As applications, these ylides were smoothly converted into α-aryl ketones after desulfurization in good yields (up to 98%) as well as in other important derivatives.