Merck
  • Exercise and environment as an intervention for neonatal alcohol effects on hippocampal adult neurogenesis and learning.
Exercise and environment as an intervention for neonatal alcohol effects on hippocampal adult neurogenesis and learning.
Neuroscience (2014-02-12)
G F Hamilton, S A Jablonski, F L Schiffino, S A St Cyr, M E Stanton, A Y Klintsova
ABSTRACT
Neonatal alcohol exposure impairs cognition and learning in adulthood and permanently damages the hippocampus. Wheel running (WR) improves hippocampus-associated learning and memory and increases the genesis and survival of newly generated neurons in the hippocampal dentate gyrus. WR significantly increases proliferation of newly generated dentate granule cells in alcohol-exposed (AE) and control rats on Postnatal Day (PD) 42 but only control rats show an increased number of surviving cells thirty days after WR (Helfer et al., 2009b). The present studies examined whether proliferation-promoting WR followed by survival-enhancing environmental complexity (EC) during adolescence could increase survival of new neurons in AE rats. On PD 4-9, pups were intubated with alcohol in a binge-like manner (5.25g/kg/day, AE), were sham-intubated (SI), or were reared normally (suckle control, SC). On PD 30 animals were assigned to WR (PD 30-42) followed by EC (PD 42-72; WR/EC) or were socially housed (SH/SH) for the duration of the experiment. All animals were injected with 200mg/kg bromodeoxyuridine (BrdU) on PD 41. In Experiment 1, survival of newly generated cells was significantly enhanced in the AE-WR/EC group in comparison with AE-SH/SH group. Experiment 2A examined trace eyeblink conditioning. In the SH/SH condition, AE impaired trace eyeblink conditioning relative to SI and SC controls. In the WR/EC condition, AE rats performed as well as controls. In Experiment 2B, the same intervention was examined using the context preexposure facilitation effect (CPFE); a hippocampus-dependent variant of contextual fear conditioning. Again, the WR/EC intervention reversed the deficit in conditioned fear to the context that was evident in the SH/SH condition. Post-weaning environmental manipulations promote cell survival and reverse learning deficits in rats that were exposed to alcohol during development. These manipulations may provide a basis for developing interventions that ameliorate learning impairments associated with human fetal alcohol spectrum disorders.
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse
Sigma-Aldrich
Anti-Glial Fibrillary Acidic Protein antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution