Merck
  • Ubiquitin-specific peptidase 2a (USP2a) deubiquitinates and stabilizes β-catenin.
Ubiquitin-specific peptidase 2a (USP2a) deubiquitinates and stabilizes β-catenin.
American journal of cancer research (2018-10-17)
Jongchan Kim, Fatemeh Alavi Naini, Yutong Sun, Li Ma
ABSTRACT
β-catenin is not only a key component of adherens junctions but also a transcriptional co-activator downstream of canonical Wnt signaling. The Wnt/β-catenin pathway plays critical roles in animal development and tissue homeostasis, while mutation or overexpression of β-catenin often leads to tumorigenesis and metastasis. Ubiquitination-mediated proteasomal degradation of β-catenin is a key molecular event in the Wnt/β-catenin pathway. Because deubiquitination of β-catenin can stabilize β-catenin and activate Wnt/β-catenin signaling, targeting the β-catenin deubiquitinase may provide a strategy for treating β-catenin-driven cancers. Here, by screening a human deubiquitinase library, we identified USP2a as a deubiquitinase that binds, deubiquitinates, and stabilizes β-catenin protein. USP2a promotes the nuclear accumulation and transcriptional activity of β-catenin, leading to elevated expression of Wnt/β-catenin target genes. Importantly, either genetic knockdown or pharmacological inhibition of USP2a leads to β-catenin destabilization. These findings suggest that USP2a may serve as a therapeutic target for targeting the cancer-promoting protein β-catenin.
Sigma-Aldrich
3X FLAG® Peptide, lyophilized powder
SKU
Pack Size
Availability
Price
Quantity
Millipore
Anti-c-Myc Agarose Affinity Gel antibody produced in rabbit, affinity isolated antibody
SKU
Pack Size
Availability
Price
Quantity
Millipore
ANTI-FLAG® antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SKU
Pack Size
Availability
Price
Quantity
Millipore
Anti-FLAG® M2 Magnetic Beads, affinity isolated antibody
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)