Merck
  • Argonaute-miRNA Complexes Silence Target mRNAs in the Nucleus of Mammalian Stem Cells.
Argonaute-miRNA Complexes Silence Target mRNAs in the Nucleus of Mammalian Stem Cells.
Molecular cell (2018-08-28)
Aishe A Sarshad, Aster H Juan, Ana Iris Correa Muler, Dimitrios G Anastasakis, Xiantao Wang, Pavol Genzor, Xuesong Feng, Pei-Fang Tsai, Hong-Wei Sun, Astrid D Haase, Vittorio Sartorelli, Markus Hafner
ABSTRACT
In mammals, gene silencing by the RNA-induced silencing complex (RISC) is a well-understood cytoplasmic posttranscriptional gene regulatory mechanism. Here, we show that embryonic stem cells (ESCs) contain high levels of nuclear AGO proteins and that in ESCs nuclear AGO protein activity allows for the onset of differentiation. In the nucleus, AGO proteins interact with core RISC components, including the TNRC6 proteins and the CCR4-NOT deadenylase complex. In contrast to cytoplasmic miRNA-mediated gene silencing that mainly operates on cis-acting elements in mRNA 3' untranslated (UTR) sequences, in the nucleus AGO binding in the coding sequence and potentially introns also contributed to post-transcriptional gene silencing. Thus, nuclear localization of AGO proteins in specific cell types leads to a previously unappreciated expansion of the miRNA-regulated transcriptome.
Sigma-Aldrich
4-Thiouridine, ≥98%
Sigma-Aldrich
ESGRO Complete Accutase, The ESGRO Complete Accutase is a cell detachment solution of proteolytic & collagenolytic enzymes, qualified for use for the detachment of mouse embryonic stem cells cultured in serum-free conditions with ESGRO Complete Clonal Grade Medium.
Sigma-Aldrich
MISSION® pLKO.1-puro Non-Target shRNA Control Plasmid DNA, Targets no known genes from any species