Merck
  • Diets supplemented with starch and corn oil, marine algae, or hydrogenated palm oil differentially modulate milk fat secretion and composition in cows and goats: A comparative study.
Diets supplemented with starch and corn oil, marine algae, or hydrogenated palm oil differentially modulate milk fat secretion and composition in cows and goats: A comparative study.
Journal of dairy science (2018-06-11)
H Fougère, C Delavaud, L Bernard
ABSTRACT
A direct comparative study of dairy cows and goats was performed to characterize the animal performance and milk fatty acid (FA) responses to 2 types of diets that induce milk fat depression in cows as well as a diet that increases milk fat content in cows but for which the effects in goats are either absent or unknown. Twelve Holstein cows and 12 Alpine goats, all multiparous, nonpregnant, and at 86 ± 24.9 and 61 ± 1.8 DIM, respectively, were allocated to 1 of 4 groups and fed diets containing no additional lipid (CTL) or diets supplemented with corn oil [5% dry matter intake (DMI)] and wheat starch (COS), marine algae powder (MAP; 1.5% DMI), or hydrogenated palm oil (HPO; 3% DMI), according to a 4 × 4 Latin square design with 28-d experimental periods. Dietary treatments had no significant effects on milk yield and DMI in both species, except for COS in cows, which decreased DMI by 17%. In cows, milk fat content was lowered by COS (-45%) and MAP (-22%) and increased by HPO (13%) compared with CTL, and in goats only MAP had an effect compared with CTL by decreasing milk fat content by 15%. In both species, COS and MAP lowered the yields (mmol/d per kg of BW) of <C16 and C16 FA. With COS, this decrease was compensated by an increase of >C16 FA in goats, but not in cows, and the >C16 FA yield decreased with MAP in both species. HPO supplementation increased the milk yield of C16 FA in cows. Compared with CTL, COS induced an increase of trans-10,cis-12 conjugated linoleic acid by 18 fold in cows and 7 fold in goats and of trans-10 18:1 by 13 fold in cows and 3 fold in goats. Moreover, other conjugated linoleic acid isomers, such as trans-10,trans-12 and trans-7,cis-9, were increased to a greater extent in cows (8 and 4 fold, respectively) compared with goats (4 and 2 fold, respectively) on the COS treatment. In both species, the responses to MAP were characterized by a decrease in the milk concentration of 18:0 (3 fold, on average) and cis-9 18:1 (2 fold, on average) combined with a 3-fold increase in the total trans 18:1, with an increase in trans-10 18:1 only observed in cows. Compared with CTL, the response to HPO was distinguished by an increase in 16:0 (10%) in cows. This comparative study clearly demonstrated that each ruminant species responds differently to COS and HPO treatments, whereas MAP caused similar effects, and that goats are less sensitive than cows to diets that induce a shift from the trans-11 toward the trans-10 ruminal pathways.
Sigma-Aldrich
Anti-14-3-3σ Antibody, clone CS112-2A8, clone CS112-2A8, Upstate®, from mouse
Sigma-Aldrich
Linoleic acid, conjugated methyl ester
Sigma-Aldrich
Linoleic acid methyl ester, cis/trans-isomers