Merck
  • An erythrosin B-based "turn on" fluorescent sensor for detecting perfluorooctane sulfonate and perfluorooctanoic acid in environmental water samples.
An erythrosin B-based "turn on" fluorescent sensor for detecting perfluorooctane sulfonate and perfluorooctanoic acid in environmental water samples.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2018-05-15)
Zhen Cheng, Lingling Du, Panpan Zhu, Qian Chen, Kejun Tan
ABSTRACT
Because of the serious harm to animals and the environment associated with perfluorooctane sulfonate (PFOS) and perfluorooctanoic acid (PFOA), a rapid, sensitive and low-cost method for detecting PFOS and PFOA is of great importance. In this paper, a novel sensing method has been proposed for the highly sensitive detection of PFOS and PFOA in environmental water samples based on the "turn-on" switch of erythrosine B (EB)-hexadecyltrimethylammonium bromide (CTAB) system. In pH 8.55 Britton-Robinson (BR) buffer, EB can react with CTAB by electrostatic attraction, resulting in a strong fluorescence quenching of EB. With a subsequent addition of the CTAB, a red-shift occurred (11 nm), followed by a significant increase in fluorescence at high surfactant concentrations. It was found that PFOS and PFOA can obviously enhance fluorescence intensity of EB-CTAB system. The enhanced fluorescence intensity is proportional to the concentration of PFOS and PFOA in the range of 0.05-10 μM with detection limit of 12.8 nM and 11.8 nM (3σ), respectively. The presented assay has been successfully applied to sensing PFOS and PFOA in real water samples with RSD ≤ 4.3% and 2.9%, respectively.
Sigma-Aldrich
Erythrosin B, Dye content ≥95 %
Sigma-Aldrich
Perfluorooctane, 98%