Merck
  • Itraconazole-Induced Inhibition on Human Esophageal Cancer Cell Growth Requires AMPK Activation.
Itraconazole-Induced Inhibition on Human Esophageal Cancer Cell Growth Requires AMPK Activation.
Molecular cancer therapeutics (2018-03-30)
Min-Bin Chen, Yuan-Yuan Liu, Zhao-Yu Xing, Zhi-Qing Zhang, Qin Jiang, Pei-Hua Lu, Cong Cao
ABSTRACT
We here evaluated the antiesophageal cancer cell activity by the antifungal drug itraconazole. Our results show that μg/mL concentrations of itraconazole potently inhibited survival and proliferation of established (TE-1 and Eca-109) and primary human esophageal cancer cells. Itraconazole activated AMPK signaling, which was required for subsequent esophageal cancer cell death. Pharmacologic AMPK inhibition, AMPKα1 shRNA, or dominant negative mutation (T172A) almost completely abolished itraconazole-induced cytotoxicity against esophageal cancer cells. Significantly, itraconazole induced AMPK-dependent autophagic cell death (but not apoptosis) in esophageal cancer cells. Furthermore, AMPK activation by itraconazole induced multiple receptor tyrosine kinases (RTKs: EGFR, PDGFRα, and PDGFRβ), lysosomal translocation, and degradation to inhibit downstream Akt activation. In vivo, itraconazole oral gavage potently inhibited Eca-109 tumor growth in SCID mice. It was yet ineffective against AMPKα1 shRNA-expressing Eca-109 tumors. The in vivo growth of the primary human esophageal cancer cells was also significantly inhibited by itraconazole administration. AMPK activation, RTK degradation, and Akt inhibition were observed in itraconazole-treated tumors. Together, itraconazole inhibits esophageal cancer cell growth via activating AMPK signaling. Mol Cancer Ther; 17(6); 1229-39. ©2018 AACR.
Sigma-Aldrich
3-Methyladenine, autophagy inhibitor
Sigma-Aldrich
Z-VAD-FMK, solid
Sigma-Aldrich
AICAR, ≥98% (HPLC), powder