Merck
  • Leptin induces SIRT1 expression through activation of NF-E2-related factor 2: Implications for obesity-associated colon carcinogenesis.
Leptin induces SIRT1 expression through activation of NF-E2-related factor 2: Implications for obesity-associated colon carcinogenesis.
Biochemical pharmacology (2018-02-11)
Na-Young Song, Yeon-Hwa Lee, Hye-Kyung Na, Jeong-Heum Baek, Young-Joon Surh
ABSTRACT
Leptin, a representative adipokine secreted from the white adipose tissue, is considered as a potential linker between obesity and cancer. SIRT1 is an NAD+-dependent histone/protein deacetylase speculated to function as an oncogene. In the present study, we found that leptin signaling-defective ob/ob and db/db mice had lower colonic expression of SIRT1 compared with leptin signaling-intact C57BL/6J mice, implying that leptin signaling is crucial for SIRT1 expression in vivo. Moreover, leptin induced up-regulation of SIRT1 in human colon cancer (HCT-116) cells. Leptin stimulated migration and invasion of cultured HCT-116 cells and tumor growth in the xenograft assay, and these effects were abrogated by a SIRT1 inhibitor sirtinol, suggesting that SIRT1 plays a role in leptin-induced colon carcinogenesis. Leptin-induced SIRT1 expression was regulated by the redox-sensitive transcription factor NF-E2-related factor 2 (Nrf2). Leptin stimulated nuclear accumulation of Nrf2 as well as its binding to the antioxidant response elements located in the SIRT1 promoter. Moreover, siRNA knockdown of Nrf2 abrogated the leptin-induced SIRT1 expression. Notably, SIRT1 was significantly reduced in colon tissues of Nrf2-null mice, lending further support to Nrf2-dependent SIRT1 expression. Expression of leptin, Nrf2 and SIRT1 was coordinately increased in human colon tumor tissues. In conclusion, leptin might play a role in colon carcinogenesis by inducing Nrf2-dependent SIRT1 overexpression.
Sigma-Aldrich
Sirtinol, ≥95% (HPLC)
Sigma-Aldrich
Amyloid Protein Non-Aβ Component, ≥80% (HPLC)