Merck
  • Immobilization of luciferase from a firefly lantern extract on glass strips as an alternative strategy for luminescent detection of ATP.
Immobilization of luciferase from a firefly lantern extract on glass strips as an alternative strategy for luminescent detection of ATP.
Journal of bioluminescence and chemiluminescence (1999-02-02)
A R Ribeiro, R M Santos, L M Rosário, M H Gil
ABSTRACT
The bioluminescent reaction catalysed by firefly luciferase has become widely established as an outstanding analytical system for assay of ATP. When used in solution, luciferase is unstable and cannot be re-used, a problem that can be partially circumvented by immobilizing the enzyme on solid substrates. Transparent glass is especially advantageous over alternative immobilizing matrices, since it allows most of the emitted photons to be detected. We report a new method for luciferase immobilization on glass which does not require prior silanization and glutaraldehyde activation, thus saving preparation time and minimizing enzyme inactivation. Our method is based on the co-immobilization by adsorption of luciferase (from a firefly lantern extract) and poly-L-lysine (PL) on non-porous glass strips. Luciferase immobilized in this way exhibits minimal variations in intersample activity, high sensitivity for ATP detection (linear luminescence responses down to 50 nmol/L) and good stability (full activity for at least 60 days when stored at -80 degrees C). PL-mediated immobilization of luciferase on glass strips provides an attractive strategy for the design of specific ATP biosensors, with potential in industry, environmental screening, medicine and biological research.
Sigma-Aldrich
Firefly Lantern Extract, powder