Merck
  • [Determination of ten pesticides of pyrazoles and pyrroles in tea by accelerated solvent extraction coupled with gas chromatography-tandem mass spectrometry].
[Determination of ten pesticides of pyrazoles and pyrroles in tea by accelerated solvent extraction coupled with gas chromatography-tandem mass spectrometry].
Se pu = Chinese journal of chromatography (2013-06-22)
Dunming Xu, Shengyu Lu, Dajie Chen, Jinchang Lan, Zhigang Zhang, Fang Yang, Yu Zhou
ABSTRACT
An effective method was developed and applied to determine the residues of ten pesticides of pyrazoles and pyrroles in tea by accelerated solvent extraction coupled with gas chromatography-tandem mass spectrometry (ASE-GC-MS/MS). The samples were extracted with ethyl acetate-hexane (1:1, v/v) for 5 min at 1.03 x 10(7) Pa and 100 degree C for one cycle. Then, they were purified by Envi-Carb/PSA column, and eluted by ethyl acetate-hexane (1:1, v/v). The analytes were determined by GC-MS/MS and quantified by external standard method. The limits of quantification were 0.003 mg/kg for fenpyroximate, 0.001 mg/kg for fipronil-sulfide, 0.002 mg/kg for fipronil, 0.005 mg/kg for fipronil-sulfone, 0.002 mg/kg for chlorfenapyr, 0.006 mg/kg for flusilazole, 0.001 mg/kg for difenzoquat, 0.001 mg/kg for pyraflufen-ethyl, 0.000 3 mg/kg for tebufenpyrad and 0.005 mg/kg for tolfenpyrad. The results show that the proposed method is sensitive and accurate for the determination of the ten pesticide residues.
Product Description
Supelco
Supel QuE, PSA/ENVI-Carb (EN) Cleanup Tube 1, pack of 100, suitable for EN 15662:2008 per BS
Supelco
Supel QuE, PSA/ENVI-Carb (EN) Cleanup Tube 2, pack of 100, suitable for EN 15662:2008 per BS
Supelco
Supel QuE, PSA/ENVI-Carb Tube 1, pk of 50, suitable for EN 15662:2008 per BS, centrifuge tube volume 15 mL , Shaker Compatible
Supelco
Supel QuE, PSA/ENVI-Carb Tube 2, pk of 50, suitable for EN 15662:2008 per BS, centrifuge tube volume 15 mL , Shaker Compatible
Supelco
Supel QuE, PSA/C18/ENVI-Carb Tube, pk of 50, AOAC® 2007.01, centrifuge tube volume 15 mL , Shaker Compatible
Supelco
Supel QuE, PSA/C18/ENVI-Carb Tube, pack of 100, AOAC® 2007.01