Merck
  • Measurement of toxic volatile organic compounds in indoor air of semiconductor foundries using multisorbent adsorption/thermal desorption coupled with gas chromatography-mass spectrometry.
Measurement of toxic volatile organic compounds in indoor air of semiconductor foundries using multisorbent adsorption/thermal desorption coupled with gas chromatography-mass spectrometry.
Journal of chromatography. A (2003-07-02)
Chien-Hou Wu, Ming-Nan Lin, Chien-Tai Feng, Kuang-Ling Yang, Yu-Shiu Lo, Jiunn-Guang Lo
ABSTRACT
A method for the qualitative and quantitative analysis of volatile organic compounds (VOCs) in the air of class-100 clean rooms at semiconductor fabrication facilities was developed. Air samples from two semiconductor factories were collected each hour on multisorbent tubes (including Carbopack B, Carbopack C, and Carbosieve SIII) with a 24-h automatic active sampling system and analyzed using adsorption/thermal desorption coupled with gas chromatography-mass spectrometry. Experimental parameters, including thermal desorption temperature, desorption time, and cryofocusing temperature, were optimized. The average recoveries and the method detection limits for the target compounds were in the range 94-101% and 0.31-0.89 ppb, respectively, under the conditions of a 1 L sampling volume and 80% relative humidity. VOCs such as acetone, isopropyl alcohol, 2-heptanone, and toluene, which are commonly used in the semiconductor and electronics industries, were detected and accurately quantified with the established method. Temporal variations of the analyte concentrations observed were attributed to the improper use of organic solvents during operation.
Product Description
Supelco
Carbopack Adsorbent, matrix Carbopack X, 60-80 mesh, bottle of 10 g
Supelco
Carbopack Adsorbent, matrix Carbopack C, 60-80 mesh, bottle of 10 g
Supelco
Carbopack Adsorbent, matrix Carbopack C, 80-100 mesh, bottle of 10 g
Supelco
Carbopack Adsorbent, matrix Carbopack X, 40-60 mesh, bottle of 10 g
Supelco
Carbopack Adsorbent, matrix Carbopack X, 120-400 mesh, bottle of 50 g
Supelco
Carbopack B, stainless steel TD tube, O.D. × L 1/4 × 3 1/2, unconditioned, pkg of 10 ea
Supelco
Carbopack B, glass TD tube, fritted, O.D. × L 1/4 in. × 3 1/2 in., preconditioned, pkg of 10 ea
Supelco
Carbopack B, glass TD tube, fritted, O.D. × L 1/4 in. × 3 1/2 in., unconditioned, pkg of 10 ea, matrix Carbopack
Supelco
Carbopack B, O.D. × L 1/4 in. × 3 1/2 in., glass TD tube, fritted, Sealed with Brass Endcaps, preconditioned, pkg of 10 ea
Supelco
FLM Carbopack X Deactivated Stainless Steel TD Tube, preconditioned, pkg of 10 tubes, O.D. × L 1/4 in. (6.35 mm) × 3 1/2 in. (89 mm)
Supelco
Carbosieve Adsorbent, matrix Carbosieve G, 60-80 mesh, bottle of 5 g
Supelco
Carbosieve Adsorbent, matrix Carbosieve S-III, 60-80 mesh, bottle of 10 g
Supelco
Carbosieve Adsorbent, matrix Carbosieve G, 80-100 mesh, bottle of 5 g
Supelco
Carbosieve SIII, stainless steel TD tube, O.D. × I.D. × L 6 mm × 4 mm × 7 in., preconditioned, pkg of 1 ea
Supelco
Carbosieve Adsorbent, matrix Carbosieve S-II, 60-80 mesh, bottle of 10 g
Supelco
Carbosieve Adsorbent, matrix Carbosieve S-II, 80-100 mesh, bottle of 10 g
Supelco
Carbopack Adsorbent, matrix Carbopack B, 60-80 mesh, bottle of 10 g
Supelco
Carbopack X, glass TD tube, fritted, O.D. × L 1/4 in. × 3 1/2 in., preconditioned, pkg of 10 ea
Supelco
2-Heptanone, analytical standard