Skip to Content
Merck
Zinc acetylacetonate hydrate 99.995% trace metals basis

Zinc acetylacetonate hydrate

Zinc acetylacetonate hydrate 99.995% trace metals basis
Synonym(s):
Bis(2,4-pentanedionato)zinc, Zn(acac)2
Linear Formula:
Zn(C5H7O2)2 · xH2O
CAS Number:
Molecular Weight:
263.61 (anhydrous basis)
EC Number:

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Chemical Composition
Markush Class
Markush Group
Reagent Type
Showing 1-2 of 2
Compare
Product Number
Description
Pricing