![Zidovudine impurity D European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/274/005/fea6028c-5ab9-47b3-a86e-109e538e1d90/640/fea6028c-5ab9-47b3-a86e-109e538e1d90.png)
Zidovudine impurity D
![Zidovudine impurity D European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/274/005/fea6028c-5ab9-47b3-a86e-109e538e1d90/640/fea6028c-5ab9-47b3-a86e-109e538e1d90.png)
Synonym(s):
Triphenylmethanol, Triphenylcarbinol, Trityl alcohol
Linear Formula:
(C6H5)3COH
CAS Number:
Molecular Weight:
260.33
Products
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing