![TSKgel® G2500HHR HPLC Column phase polystyrene-divinylbenzene, L × I.D. 30 cm × 7.8 mm, 5 μm particle size](/deepweb/assets/sigmaaldrich/product/images/380/683/7329afd5-92b6-4974-b402-9c8aa3e087dd/640/7329afd5-92b6-4974-b402-9c8aa3e087dd.jpg)
TSKgel® G2500HHR (5 µm) HPLC Columns
![TSKgel® G2500HHR HPLC Column phase polystyrene-divinylbenzene, L × I.D. 30 cm × 7.8 mm, 5 μm particle size](/deepweb/assets/sigmaaldrich/product/images/380/683/7329afd5-92b6-4974-b402-9c8aa3e087dd/640/7329afd5-92b6-4974-b402-9c8aa3e087dd.jpg)
Synonym(s):
TSKgel® HHR Column
Products
Product Category
Manufacturer
Product Line
Agency
USP Suitability
Separation Technique
ID (mm)
Length (cm)
Particle Size (µm)
Pore Size (Å)
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing