Skip to Content
Merck
Triethoxymethylsilane 99%

Triethoxymethylsilane

Triethoxymethylsilane 99%
Synonym(s):
Methyltriethoxysilane, Triethoxymethylsilane
Linear Formula:
(C2H5O)3SiCH3
CAS Number:
Molecular Weight:
178.30

Products

Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing