
Triclabendazole for system suitability

Synonym(s):
Triclabendazole, 5-Chloro-6-(2,3-dichlorophenoxy)-2-(methylthio)-1H-benzimidazole
Empirical Formula (Hill Notation):
C14H9Cl3N2OS
CAS Number:
Molecular Weight:
359.66
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing