Skip to Content
Merck
vibrant-m

Tranexamic acid impurity C

vibrant-m
Synonym(s):
(RS)-4-(Aminomethyl)cyclohex-1-enecarboxylic acid
Empirical Formula (Hill Notation):
C8H13NO2
CAS Number:
Molecular Weight:
155.19

Products

Formula Weight
Manufacturer
Quality Segment
Application
API Family
Reference Material Type
Showing 1-1 of 1