TOYOPEARL® AF-rProtein A-650F Bulk Media
Synonym(s):
TOYOPEARL® AF-rProtein A-650F Bulk Media
Products
Manufacturer
Physical Form
Product Line
Separation Technique
Package Size
Particle Size (µm)
Pore Size (Å)
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing