Skip to Content
Merck
Toyocamycin ≥98% (HPLC), from Streptomyces rimosus

Toyocamycin

Toyocamycin ≥98% (HPLC), from Streptomyces rimosus
Synonym(s):
4-Aminopyrrolo[2,3-d]pyrimidine-5-carbonitrile 7-(β-D-ribofuranoside), 7-Deaza-7-cyanoadenosine, NSC 63701, NSC 99843, Neuro 000027, Unamycin B, Vengicide
Empirical Formula (Hill Notation):
C12H13N5O4
CAS Number:
Molecular Weight:
291.26

Products

Product Category
Brand
Biological Source
Formula Weight
Physical Form
Purity
Activity Spectrum
Research Area
Quality Segment
Mode of Action
Showing 1-1 of 1
Compare
Product Number
Description
Pricing