Torasemide anhydrous
Synonym(s):
Torsemide, Demadex, N-[[(1-Methylethyl)amino]carbonyl]-4-[(3-methylphenyl)amino]-3-pyridinesulfonamide
Empirical Formula (Hill Notation):
C16H20N4O3S
CAS Number:
Molecular Weight:
348.42
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing