![Tolfenpyrad PESTANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/216/309/c3ea9953-2ea5-4e39-8d95-85053646c071/640/c3ea9953-2ea5-4e39-8d95-85053646c071.png)
Tolfenpyrad
![Tolfenpyrad PESTANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/216/309/c3ea9953-2ea5-4e39-8d95-85053646c071/640/c3ea9953-2ea5-4e39-8d95-85053646c071.png)
Synonym(s):
4-Chloro-3-ethyl-1-methyl-N-[4-(p-tolyloxy)benzyl]pyrazole-5-carboxamide, 4-Chloro-3-ethyl-1-methyl-N-{[4-(4-methylphenoxy)phenyl]methyl}-1H-pyrazole-5-carboxamide
Empirical Formula (Hill Notation):
C21H22ClN3O2
CAS Number:
Molecular Weight:
383.87
Products
Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing