![Titriplex® III (ethylene dinitrilo tetraacetic acid disodium salt dihydrate), EMPROVE® EXPERT, Ph. Eur., BP, ChP, JP, USP](/deepweb/assets/sigmaaldrich/product/structures/177/279/de9a18cd-88da-498c-ad1f-ca6a67d22fab/640/de9a18cd-88da-498c-ad1f-ca6a67d22fab.png)
Titriplex® III
![Titriplex® III (ethylene dinitrilo tetraacetic acid disodium salt dihydrate), EMPROVE® EXPERT, Ph. Eur., BP, ChP, JP, USP](/deepweb/assets/sigmaaldrich/product/structures/177/279/de9a18cd-88da-498c-ad1f-ca6a67d22fab/640/de9a18cd-88da-498c-ad1f-ca6a67d22fab.png)
Synonym(s):
Ethylenediaminetetraacetic acid disodium salt dihydrate, Disodium ethylenediaminetetraacetate dihydrate, EDTA disodium salt, EDTA-Na2, Edathamil, Edetate disodium salt dihydrate, Sequestrene Na2
Empirical Formula (Hill Notation):
C10H14N2Na2O8 · 2H2O
CAS Number:
Molecular Weight:
372.24
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Agency
Quality Segment
Application
Reaction Type
Special Grade
Technique
Showing 1-2 of 2