Skip to Content
Merck
Timolol Related Compound B certified reference material, pharmaceutical secondary standard

Timolol Related Compound B

Timolol Related Compound B certified reference material, pharmaceutical secondary standard
Synonym(s):
3-(tert-Butylamino)-2-(4-morpholino-1,2,5-thiadiazol-3-yloxy)propan-1-ol, Isotimolol
Empirical Formula (Hill Notation):
C13H24N4O3S
CAS Number:
Molecular Weight:
316.42

Products

Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade