Skip to Content
Merck
vibrant-m

Ticlopidine Related Compound A

vibrant-m
Synonym(s):
4-Oxo-4,5,6,7-tetrahydrothieno[3,2-c]pyridine
Empirical Formula (Hill Notation):
C7H7NOS
CAS Number:
Molecular Weight:
153.20

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1